Funnel holders
1 funnel
Catalog # 221-1261
Supplier: KARTELL
Jump to:
Product Details & Documents
PP, white, unbreakable.
- For funnels 50 to 120 mm Ø
- With bosshead to fit rods 8 to 14 mm Ø
- Interchangeable insert for different sizes
Specifications
- For:1 funnel
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



