Microplates, 96-well
96-well plate, U-bottom, aseptic, Clear, 367 µl, 96
Catalog # KART2621
Supplier: KARTELL
Jump to:
Product Details & Documents
Multiple-well plates (96 sample wells) in clear PS. Available in three shapes: flat bottom, U bottom and V bottom wells.
- Aseptic or sterile
- Single-wrapped
Ordering information: PS covers are sold separately.
Specifications
- No. of wells:96
- Volume:367 µl
- Colour:Clear
- Description:96-well plate, U-bottom, aseptic
Accessories
Product Information
- Catalog #AvailabilityPrice
- Lids for 96 well microtitre plates, PS, asepticLids for 96 well microtitre plates, PS, asepticKART2623Log in for availability1 * 50 ST - 83,30 €
Product Information
- Catalog #AvailabilityPrice
- Lids for 96 well microtitre plates, PS, sterileLids for 96 well microtitre plates, PS, sterileKART2623/SLog in for availability1 * 50 ST - 87,00 €