Orders LinkContactUsLinkComponent
Sample Cups for Analysers
Sample Cups for Analysers
Special cross cut cap for Centrifichem® and Gemsaec® analyser cups
Catalog # 391-1019
Supplier:   KARTELL
Sample Cups for Analysers
Catalog # 391-1019
Supplier:  KARTELL
Sample Cups for Analysers
Sample Cups for Analysers
Special cross cut cap for Centrifichem® and Gemsaec® analyser cups
Price
33,60 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
PS, transparent.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Description:
    Special cross cut cap for Centrifichem® and Gemsaec® analyser cups
  • Seq:
    09