Order Entry
Belgium
Orders LinkContactUsLinkComponent
Volumetric flasks, class B

Volumetric flasks, class B

250 ml, 19/26, 0,30 ± ml
Catalog # 612-3008
Supplier:   KARTELL
Volumetric flasks, class B
Catalog # 612-3008
Supplier:  KARTELL
Volumetric flasks, class B
Volumetric flasks, class B

250 ml, 19/26, 0,30 ± ml

Price
42,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PMP (TPX®), transparent, with stopper.

  • Autoclavable
  • For working temperatures up to +170 °C
  • With NS stoppers
Specifications
  • NS:
    19/26
  • Capacity:
    250 ml
  • Tolerance:
    0,30 ± ml