Order Entry
Belgium
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

280 mm, 310 mm
Catalog # 221-0081
Supplier:   KARTELL
Funnels with short stem
Catalog # 221-0081
Supplier:  KARTELL
Funnels with short stem
Funnels with short stem

280 mm, 310 mm

Price
22,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

HDPE, translucent.

  • Good chemical resistance
  • With raised lip to prevent overflowing
  • External ribs for unobstructed removal of air when filtering

With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

Documents

Specifications
  • Funnel O-Ø:
    310 mm
  • Length:
    280 mm
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.