Order Entry
Belgium
Orders LinkContactUsLinkComponent
Funnels with short stem

Funnels with short stem

154 mm, 140 mm
Catalog # 221-1523
Supplier:   KARTELL
Funnels with short stem
Catalog # 221-1523
Supplier:  KARTELL
Funnels with short stem
Funnels with short stem

154 mm, 140 mm

Price
6,10 €
Log in for availability
1 * 1 ST
Product Details & Documents

HDPE, translucent.

  • Good chemical resistance
  • With raised lip to prevent overflowing
  • External ribs for unobstructed removal of air when filtering

With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.

Documents

Specifications
  • Funnel O-Ø:
    140 mm
  • Length:
    154 mm
  • Seq:
    04
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.