Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
100 mm, 100 mm
Catalog # 221-1521
Supplier:   KARTELL
Funnels with short stem
Catalog # 221-1521
Supplier:  KARTELL
Funnels with short stem
Funnels with short stem
100 mm, 100 mm
Price
4,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
HDPE, translucent.
  • Good chemical resistance
  • With raised lip to prevent overflowing
  • External ribs for unobstructed removal of air when filtering
With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses. Suspension eye for easy storage.
Documents
Specifications
  • Funnel O-Ø:
    100 mm
  • Length:
    100 mm
  • Seq:
    02