Order Entry
Belgium
Orders LinkContactUsLinkComponent
Accessories for fluid and solid samplers

Accessories for fluid and solid samplers

Cleaning brush, PVC/stainless steel, Ø: 25 mm, 600 mm
Catalog # 331-2173
Supplier:   BURKLE
Accessories for fluid and solid samplers
Catalog # 331-2173
Supplier:  BURKLE
Accessories for fluid and solid samplers
Accessories for fluid and solid samplers

Cleaning brush, PVC/stainless steel, Ø: 25 mm, 600 mm

Price
80,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cleaning brush, PVC/stainless steel, Ø: 25 mm, 600 mm
  • Seq:
    3