Order Entry
Belgium
Orders LinkContactUsLinkComponent
Scoops

Scoops

2000 ml, 370 mm
Catalog # 231-0206
Supplier:   BOCHEM
Scoops
Catalog # 231-0206
Supplier:  BOCHEM
Scoops
Scoops

2000 ml, 370 mm

Price
41,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.

  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling, anti-magnetic
Specifications
  • Capacity:
    2000 ml
  • Length:
    370 mm