Order Entry
Belgium
Orders LinkContactUsLinkComponent
VWR®, pH buffer capsules

VWR®, pH buffer capsules

Capsules, 4,01, ±0,02, Orange, 100 ml, Plastic tube, 5 plastic tubes, 10 capsules/tube, 50
Catalog # 332732B
Supplier:   VWR Chemicals
VWR®, pH buffer capsules
Catalog # 332732B
Supplier:  VWR Chemicals
CAS Number:  
VWR®, pH buffer capsules
VWR®, pH buffer capsules

Capsules, 4,01, ±0,02, Orange, 100 ml, Plastic tube, 5 plastic tubes, 10 capsules/tube, 50

CAS Number:  
Price
216,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
  • Colour coded for easy identification
  • Preservative-free
  • Extended shelf life
  • Add one capsule to 100 ml of distilled water
Specifications

Specifications

  • Pack type:
    Plastic tube
  • Pk:
    50
  • Colour:
    Orange
  • Description:
    Capsules
  • pH accuracy:
    ±0,02
  • Final volume:
    100 ml
  • Pk Info:
    5 plastic tubes, 10 capsules/tube
  • pH value:
    4,01

Specifications