Order Entry
Belgium
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rod, Glass, 6 - 7 mm, 150 mm
Catalog # 441-0266
Supplier:   EMMANUEL PIERRE
Stirring rods
Catalog # 441-0266
Supplier:  EMMANUEL PIERRE
Stirring rods
Stirring rods

Stirring rod, Glass, 6 - 7 mm, 150 mm

Price
Inquire For Price
Log in for availability
1 * 10 ST
Product Details & Documents

Made from ordinary glass.

Specifications
  • Ø:
    6 - 7 mm
  • Description:
    Stirring rod
  • Length:
    150 mm
  • Material:
    Glass