Order Entry
Belgium
Orders LinkContactUsLinkComponent
Quinoline 99%

Quinoline 99%

Glass bottle for solids, 100 g
Catalog # ACRO221141000
Toxic
Health
Environment
Quinoline 99%
Catalog # ACRO221141000
CAS Number:  91-22-5
Quinoline 99%
Quinoline 99%

Glass bottle for solids, 100 g

CAS Number:  91-22-5
Price
54,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:13,08160
  • Boiling Pt:237 °C (760 mmHg)
  • Melting Pt:–15 °C
  • Density:1.095 g/cm³
  • Flash Pt:101 °C
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to dark brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6230 to 1.6270 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications