Sodium deoxycholate 99%, extra pure
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Sodium deoxycholate 99%, extra pure
Catalog # ACRO218591000
Supplier: Thermo Fisher Scientific
CAS Number: 302-95-4
Jump to:
Product Details & Documents
Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
- CAS Number:302-95-4
- Formula:C₂₄H₃₉NaO₄
- MDL Number:MFCD00064139
- Molecular Weight:414.56 g/mol
- Melting Pt:357…365 °C
- Storage Temperature:Ambient
- EINECS:206-132-7
Specifications
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 % (2 g, 105°C, 4 h)
- TLC:≤2 % Sodium cholate
- pH:7.5 to 10 (5 % in water)
- Specific optical rotation:+42° to +46° (20°C, 589 nm) (c=2, water)
- Titration with HClO4:98.5 to 102.0 % (on dry substance)
- Heavy metals (as Pb):≤20 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



