Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sodium deoxycholate 99%, extra pure

Sodium deoxycholate 99%, extra pure

Glass bottle for solids, 25 g
Catalog # ACRO218590250
Irritant
Sodium deoxycholate 99%, extra pure
Catalog # ACRO218590250
CAS Number:  302-95-4
Sodium deoxycholate 99%, extra pure
Sodium deoxycholate 99%, extra pure

Glass bottle for solids, 25 g

CAS Number:  302-95-4
Price
52,30 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate
  • CAS Number:302-95-4
  • Formula:C₂₄H₃₉NaO₄
  • MDL Number:MFCD00064139
  • Molecular Weight:414.56 g/mol
  • Melting Pt:357…365 °C
  • Storage Temperature:Ambient
  • EINECS:206-132-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 % (2 g, 105°C, 4 h)
  • TLC:
    ≤2 % Sodium cholate
  • pH:
    7.5 to 10 (5 % in water)
  • Specific optical rotation:
    +42° to +46° (20°C, 589 nm) (c=2, water)
  • Titration with HClO4:
    98.5 to 102.0 % (on dry substance)
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.