Micro pipettes, disposable, Microcaps®
3,0 µl, 32 mm
Catalog # DRUM1-000-0030
Supplier: DRUMMOND
Jump to:
Product Details & Documents
Micro pipettes are precision-bore glass capillary tubes cut to predetermined lengths so that fill volumes are constant and precise. Capillary action draws the fluid into the tube and a squeeze of the bulb dispenses it with accuracy of ±1%.
For fast, safe, clean transfer of liquids in micro quantities. Suitable for both general and hazardous microchemical applications.
Ordering information: Delivery includes 100 pipettes and one bulb dispenser.
Specifications
- Capacity:3,0 µl
- Length:32 mm
- Seq:09
Accessories
Product Information
- Catalog #AvailabilityPrice
- Silicone tips for Microcaps®
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



