Orders LinkContactUsLinkComponent
Test sets
Test sets
Test set III for organic vapours
Catalog # 151-0179
Supplier:   Dräger
Test sets
Catalog # 151-0179
Supplier:  Dräger
Test sets
Test sets
Test set III for organic vapours
Price
112,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Specifications
  • Description:
    Test set III for organic vapours
  • Seq:
    03
  • Cat. No.:
    151-0179
  • Model:
    Test set III for organic vapours
  • Measurement time:
    120 s