Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanadium(V) oxide 99.6+%

Vanadium(V) oxide 99.6+%

Glass bottle for solids, 250 g
Catalog # ACRO206422500
Corrosive
Irritant
Health
Environment
Vanadium(V) oxide 99.6+%
Catalog # ACRO206422500
CAS Number:  1314-62-1
Vanadium(V) oxide 99.6+%
Vanadium(V) oxide 99.6+%

Glass bottle for solids, 250 g

CAS Number:  1314-62-1
Price
93,50 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Vanadium pentoxide, di-Vanadium pentaoxide
  • CAS Number:1314-62-1
  • Formula:O₅V₂
  • MDL Number:MFCD00011457
  • Molecular Weight:181.88 g/mol
  • Merck Index:13,09987
  • Boiling Pt:1750 °C (760 mmHg)
  • Melting Pt:690 °C
  • Density:3.35 g/cm³
  • Storage Temperature:Ambient
  • EINECS:215-239-8
  • UN:2862
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to orange-brown Powder
  • Assay:
    ≥99.60 %
  • Chromium (Cr):
    ≤100 ppm
  • Iron (Fe):
    ≤300 ppm
  • Potassium (K):
    ≤200 ppm
  • Silicon (Si):
    ≤200 ppm
  • Sodium (Na):
    ≤200 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications