Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Chlorophenol 99+%

4-Chlorophenol 99+%

Glass bottle for solids, 500 g
Catalog # ACRO181005000
Irritant
Environment
4-Chlorophenol 99+%
Catalog # ACRO181005000
CAS Number:  106-48-9
4-Chlorophenol 99+%
4-Chlorophenol 99+%

Glass bottle for solids, 500 g

CAS Number:  106-48-9
Price
65,10 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Merck Index:13,02173
  • Boiling Pt:220 °C (760 mmHg)
  • Melting Pt:41…45 °C
  • Density:1.26 g/cm³
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light brown Crystals or fused mass
  • Infrared spectrum:
    Conforms
  • GC:
    ≥99.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.