4-Chlorophénol 99+%
Flacon verre pour solides, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: p-Chlorophenol
- CAS Number:106-48-9
- Formula:C₆H₅ClO
- MDL Number:MFCD00002318
- Molecular Weight:128.56 g/mol
- Merck Index:13,02173
- Boiling Pt:220 °C (760 mmHg)
- Melting Pt:41…45 °C
- Density:1,26 g/cm³
- Flash Pt:102 °C
- Storage Temperature:Température ambiante
- EINECS:203-402-6
- UN:2020
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:White to light brown Crystals or fused mass
- Infrared spectrum:Conforms
- GC:≥99.0 %
Specifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:100 g



