Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cyclohexane 99.5% for analysis

Cyclohexane 99.5% for analysis

Glass bottle, 2,5 L
Catalog # 176810025.
Irritant
Health
Environment
Flammable
Cyclohexane 99.5% for analysis
Catalog # 176810025.
CAS Number:  110-82-7
Cyclohexane 99.5% for analysis
Cyclohexane 99.5% for analysis

Glass bottle, 2,5 L

CAS Number:  110-82-7
Price
153,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:6.5 °C
  • Density:0.77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Ambient
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %
  • Substance darkened by H2SO4:
    ≤10 APHA
  • Calcium (Ca):
    ≤0.5 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Potassium (K):
    ≤0.2 ppm
  • Sodium (Na):
    ≤1 ppm
  • Zinc (Zn):
    ≤0.1 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.