Acetone 99.5+% for analysis
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Acetone 99.5+% for analysis
Catalog # ACRO176805000
Supplier: Thermo Fisher Scientific
CAS Number: 67-64-1
Aller à :
Product Details & Documents
Synonymes: 2-Propanone, DMK, Propanone, Dimethyl ketone
- Numéro CAS:67-64-1
- Formule:CH₃COCH₃
- Numéro MDL:MFCD00008765
- Poids moléculaire:58.08 g/mol
- Index Merck:13,00067
- Point d'ébullition:56 °C (760 mmHg)
- Point de fusion:–95 °C
- Densité:0.79 g/cm³
- Point d'éclair:–20 °C
- Température de stockage:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Spécifications
Résultats des tests de spécification
- Appearance:Clear liquid
- Water:≤0.5 % (Coulometric)
- GC:≥99.50 %
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0005 meq/g
- Cadmium (Cd):≤0.05 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.05 ppm
- KMnO4-reducing substance:≤5 ppm
Spécifications
- Pack type:Glass bottle
- Pk:500 mL
- Cat. No.:ACRO176805000
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



