Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Acetone 99.5+% for analysis

Glass bottle, 2,5 L
Catalog # ACRO176800025
Irritant
Flammable
   Safer Human and Environmental Health
Acetone 99.5+% for analysis
Catalog # ACRO176800025
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone 99.5+% for analysis
Acetone 99.5+% for analysis

Glass bottle, 2,5 L

CAS Number:  67-64-1
Price
91,80 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56 °C (760 mmHg)
  • Melting Pt:–95 °C
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.50 %
  • Color scale:
    ≤5 APHA
  • Residue after evaporation:
    ≤0.001 %
  • Acidity/alkalinity:
    ≤0.0005 meq/g
  • Cadmium (Cd):
    ≤0.05 ppm
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.5 ppm
  • Lead (Pb):
    ≤0.1 ppm
  • Magnesium (Mg):
    ≤0.1 ppm
  • Zinc (Zn):
    ≤0.05 ppm
  • KMnO4-reducing substance:
    ≤5 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Cat. No.:
    ACRO176800025
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications