Acetone 99.5+% for analysis
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Acetone 99.5+% for analysis
Catalog # ACRO176800010
Supplier: Thermo Fisher Scientific
CAS Number: 67-64-1
Jump to:
Product Details & Documents
Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone
- CAS Number:67-64-1
- Formula:CH₃COCH₃
- MDL Number:MFCD00008765
- Molecular Weight:58.08 g/mol
- Merck Index:13,00067
- Boiling Pt:56 °C (760 mmHg)
- Melting Pt:–95 °C
- Density:0.79 g/cm³
- Flash Pt:–20 °C
- Storage Temperature:Ambient
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.5 % (Coulometric)
- GC:≥99.50 %
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0005 meq/g
- Cadmium (Cd):≤0.05 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.5 ppm
- Lead (Pb):≤0.1 ppm
- Magnesium (Mg):≤0.1 ppm
- Zinc (Zn):≤0.05 ppm
- KMnO4-reducing substance:≤5 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:ACRO176800010
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



