CRYO-VIAL BOXES 1.0/1.5ML 100 VIALS/BOX 1 * 10 items
Catalog # BARNCS509X24
Supplier: Thermo Fisher Scientific
CRYO-VIAL BOXES 1.0/1.5ML 100 VIALS/BOX 1 * 10 items
Catalog # BARNCS509X24
Supplier: Thermo Fisher Scientific
Frequently Bought Together
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
ANALYTICAL SALES Stackable Tray with 8×30 Shell VialsC ACI
Supplier: MARKET SOURCE PART PROCESS
Sample container with snap-on cap
Supplier: VITLAB
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier: VWR Collection
pluriStrainer® 20 µm (Zellsieb) 1 * 25 items
Supplier: MARKET SOURCE PART PROCESS
Freezer boxes (Cryoboxes), True North®
Supplier: HEATHROW
Roller bottle standard, surface treated, 1000 ml vent cap 1 * 24 items
Supplier: GUANGZHOU JET BIOFILTRATION
Sterilin® Dippas® Milk Dippers
Supplier: STERILIN
Sample containers with screw cap
Supplier: VITLAB
Storage Box for Sample Tubes
Supplier: Simport Scientific
VWR®, Polystyrene Assay Plates, 96- and 384-Well
Supplier: VWR Collection
Storage microplates 384-well polypropylene, Corning®
Supplier: Corning
Multidishes, Nunc™ UpCell™ surface
Supplier: Thermo Fisher Scientific
Accessories and consumables for KingFisher™ Flex
Supplier: Thermo Fisher Scientific
Cryobox B68 1 * 1 items
Supplier: GLW