Mesitylene 97%, pure
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,3,5-Trimethylbenzene
- CAS Number:108-67-8
- Formula:C₉H₁₂
- MDL Number:MFCD00008538
- Molecular Weight:120.19 g/mol
- Merck Index:13,05934
- Boiling Pt:163…166 °C (760 mmHg)
- Melting Pt:–45 °C
- Density:0.8675 g/cm³
- Flash Pt:44 °C
- Storage Temperature:Ambient
- EINECS:203-604-4
- UN:2325
- ADR:3,III
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4980 to 1.5000 (20°C, 589 nm)
- GC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:1 L



