Orders LinkContactUsLinkComponent
Pipette tips, microvolume, Corning®
Round tip, 0,1 - 10 µl, -, Transparent, Gilson, other Ultra-Micropipettors, Bulk; 10000 tips/bag
  613-4245
   Low Manufacturing Impact
 :   Corning
Pipette tips, microvolume, Corning®
  613-4245
 :  Corning
   Low Manufacturing Impact
Pipette tips, microvolume, Corning®
Pipette tips, microvolume, Corning®
Round tip, 0,1 - 10 µl, -, Transparent, Gilson, other Ultra-Micropipettors, Bulk; 10000 tips/bag
496,00 €
1 * 10.000 ST
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
  • All racked tips are certified RNase-/DNase-free and non pyrogenic
  • Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
  • Packed:
    Bulk; 10000 tips/bag
  • Sterile:
    -
  • Volume:
    0,1 - 10 µl
  • Seq:
    02
  • Tip style:
    Round tip
  • Tip colour:
    Transparent
  • For use with:
    Gilson, other Ultra-Micropipettors
  • Cat. No.:
    613-4245
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact