Corning® Microplate Lids
Product Details & Documents
All lids are made of rigid polystyrene except where indicated.
All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning® microplates.
The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
Specifications
- Sterility:Sterile
- Description:Low evaporation lid with corner notch and condensation rings for 96-well microplates only (not 2 ml block)
- Cat. No.:734-1669
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



