Order Entry
Belgium
Orders LinkContactUsLinkComponent
L(-)-Proline 99+%

L(-)-Proline 99+%

Glass bottle for solids, 25 g
Catalog # ACRO157620250
L(-)-Proline 99+%
Catalog # ACRO157620250
CAS Number:  147-85-3
L(-)-Proline 99+%
L(-)-Proline 99+%

Glass bottle for solids, 25 g

CAS Number:  147-85-3
Price
60,60 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
  • CAS Number:147-85-3
  • Formula:C₅H₉NO₂
  • MDL Number:MFCD00064318
  • Molecular Weight:115.13 g/mol
  • Merck Index:13,07871
  • Boiling Pt:245 °C (1013 hPa)
  • Melting Pt:228…233 °C
  • Density:1.35 g/cm³
  • Storage Temperature:Ambient
  • EINECS:205-702-2

Documents

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.3 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.1 %
  • UV transmittance:
    ≥95 % (c=10, H2O, 430 nm) 1 cm cell
  • pH:
    5.5 to 7 (2.5 % at 25°C)
  • Specific optical rotation:
    -84.5° to -86.8° (20°C, 589 nm) (c=4, H2O)
  • Titration with HClO4:
    99.0 to 101.0 %
  • Chloride (Cl):
    ≤200 ppm
  • Sulfate (SO4):
    ≤200 ppm
  • Ammonium (NH4):
    ≤200 ppm
  • Arsenic (As):
    ≤1 ppm
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Other amino acids:
    ≤0.5 % (TLC)
  • Insoluble matter:
    ≤0.005 % (in water)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.