4-Nitrophenol 99%
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Hydroxy-4-nitrobenzene, p-Nitrophenol
- CAS Number:100-02-7
- Formula:C₆H₅NO₃
- MDL Number:MFCD00007331
- Molecular Weight:139.11 g/mol
- Merck Index:13,06654
- Boiling Pt:279 °C (760 mmHg)
- Melting Pt:>112 °C
- Density:1,48 g/cm³ (20 °C)
- Flash Pt:169 °C
- Storage Temperature:Ambient
- EINECS:202-811-7
- UN:1663
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:Yellow to brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:112 to 115 °C
- GC:≥98.5 %
- Titration with NaOH:≥96.0 to ≤104.0 %
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Cat. No.:ACRO157051000
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



