Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Nitrophenol 99%

4-Nitrophenol 99%

Plastic bottle, 1 kg
Catalog # ACRO157050010
Toxic
Health
4-Nitrophenol 99%
Catalog # ACRO157050010
CAS Number:  100-02-7
4-Nitrophenol 99%
4-Nitrophenol 99%

Plastic bottle, 1 kg

CAS Number:  100-02-7
Price
118,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1-Hydroxy-4-nitrobenzene, p-Nitrophenol
  • CAS Number:100-02-7
  • Formula:C₆H₅NO₃
  • MDL Number:MFCD00007331
  • Molecular Weight:139.11 g/mol
  • Merck Index:13,06654
  • Boiling Pt:279 °C (760 mmHg)
  • Melting Pt:>112 °C
  • Density:1,48 g/cm³ (20 °C)
  • Flash Pt:169 °C
  • Storage Temperature:Ambient
  • EINECS:202-811-7
  • UN:1663
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    112 to 115 °C
  • GC:
    ≥98.5 %
  • Titration with NaOH:
    ≥96.0 to ≤104.0 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications