Orders LinkContactUsLinkComponent
Thermometer test caps
Thermometer test caps
TX24L, test cap, Lumberg type, +3 °C
Catalog # 620-0378
Supplier:   COMARK
Thermometer test caps
Catalog # 620-0378
Supplier:  COMARK
Thermometer test caps
Thermometer test caps
TX24L, test cap, Lumberg type, +3 °C
Price
156,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
Specifications
  • Range:
    +3 °C
  • Description:
    TX24L, test cap, Lumberg type