Thermometer test caps
TX23L, test cap, Lumberg type, 0 °C
620-0377
: COMARK
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
- Range:0 °C
- Description:TX23L, test cap, Lumberg type