Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Galactose 99+%

Plastic bottle, 500 g
Catalog # ACRO150615000
D-(+)-Galactose 99+%
Catalog # ACRO150615000
CAS Number:  59-23-4
D-(+)-Galactose 99+%
D-(+)-Galactose 99+%

Plastic bottle, 500 g

CAS Number:  59-23-4
Price
238,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:165…168 °C
  • Density:1,5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 % (K.F.)
  • Sulfated ash:
    ≤0.1 % (on dry substance)
  • Solubility:
    (10 % in water) Clear colorless
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    +78° to +81.5° (20°C, 589 nm) (c=10, water, NH3) (after 30 minutes, on dry substance)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications