Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Galactose 99+%

D-(+)-Galactose 99+%

Plastic bottle, 100 g
Catalog # ACRO150611000
D-(+)-Galactose 99+%
Catalog # ACRO150611000
CAS Number:  59-23-4
D-(+)-Galactose 99+%
D-(+)-Galactose 99+%

Plastic bottle, 100 g

CAS Number:  59-23-4
Price
68,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:165…168 °C
  • Density:1,5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 % (K.F.)
  • Sulfated ash:
    ≤0.1 % (on dry substance)
  • Solubility:
    (10 % in water) Clear colorless
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    +78° to +81.5° (20°C, 589 nm) (c=10, water, NH3) (after 30 minutes, on dry substance)

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.