Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Mannose 99+%

D-(+)-Mannose 99+%

Plastic bottle, 1 kg
Catalog # ACRO150600010
D-(+)-Mannose 99+%
Catalog # ACRO150600010
CAS Number:  3458-28-4
D-(+)-Mannose 99+%
D-(+)-Mannose 99+%

Plastic bottle, 1 kg

CAS Number:  3458-28-4
Price
875,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1,539 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White Powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.45 %
  • HPLC:
    ≥99.0 %
  • Appearance of solution:
    (10 % in water) Clear
  • Bacterial endotoxins:
    (LAL-test) 0.5 E.U. in 1 ml of a 0.1% soln.
  • Specific optical rotation:
    +13.5° to +14.9° (20°C, 589 nm) (c=10, water) (after 24 hours, on dry substance)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.