Order Entry
Belgium
Orders LinkContactUsLinkComponent
Vanillin 99%, pure

Vanillin 99%, pure

Plastic bottle, 250 g
Catalog # ACRO140822500
Irritant
Vanillin 99%, pure
Catalog # ACRO140822500
CAS Number:  121-33-5
Vanillin 99%, pure
Vanillin 99%, pure

Plastic bottle, 250 g

CAS Number:  121-33-5
Price
97,70 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:170 °C (15 mmHg)
  • Melting Pt:81…84 °C
  • Density:1,056 g/cm³ (20 °C)
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    81 to 84 °C
  • Titration with NaOH:
    ≥98.5 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.