Orders LinkContactUsLinkComponent
Flat bottom chambers
Flat bottom chambers
Chamber for 200×200 mm plates, without lid
Catalog # 552-0022
Supplier:   CAMAG
Flat bottom chambers
Catalog # 552-0022
Supplier:  CAMAG
Flat bottom chambers
Flat bottom chambers
Chamber for 200×200 mm plates, without lid
Price
329,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These flat bottom chamber allow the chromatogram to be developed in a tank atmosphere of partial or complete saturation with solvent vapours. The degree of layer pre-saturation cannot be controlled unless additional accessories are used.
Specifications
  • Description:
    Chamber for 200×200 mm plates, without lid
  • Seq:
    06
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Saturation pads
    Saturation pads
    552-0079
    Log in for availability
    1 * 100 ST - 245,00 €