Orders LinkContactUsLinkComponent
Twin trough chambers
Twin trough chambers
Chamber for 200×200 mm plates, with glass lid
Catalog # 552-0020
Supplier:   CAMAG
Twin trough chambers
Catalog # 552-0020
Supplier:  CAMAG
Twin trough chambers
Twin trough chambers
Chamber for 200×200 mm plates, with glass lid
Price
386,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Twin-trough chambers offer several ways to improve the results of TLC/HPTLC developing techniques.
  • Low solvent consumption
  • Reproducible pre-equilibration of the plate with solvent vapour is possible by placing it in the empty trough opposite that containing the solvent
Development only starts when the solvent is introduced into the trough with the plate.
* ADC: automatic developing chamber.
Specifications
  • Description:
    Chamber for 200×200 mm plates, with glass lid
  • Seq:
    05