Orders LinkContactUsLinkComponent
Lids for flat bottom chambers
Lids for flat bottom chambers
Glass lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
Catalog # 552-0017
Supplier:   CAMAG
Lids for flat bottom chambers
Catalog # 552-0017
Supplier:  CAMAG
Lids for flat bottom chambers
Lids for flat bottom chambers
Glass lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
Price
20,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Glass lid for 200×200 mm and 200×100 mm twin trough and flat bottom chambers
  • Seq:
    09