Orders LinkContactUsLinkComponent
Lids for flat bottom chambers
Lids for flat bottom chambers
Stainless steel lid for 100×100 mm twin trough and flat bottom chambers
Catalog # 552-0008
Supplier:   CAMAG
Lids for flat bottom chambers
Catalog # 552-0008
Supplier:  CAMAG
Lids for flat bottom chambers
Lids for flat bottom chambers
Stainless steel lid for 100×100 mm twin trough and flat bottom chambers
Price
36,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Stainless steel lid for 100×100 mm twin trough and flat bottom chambers
  • Seq:
    10