Order Entry
Belgium
Orders LinkContactUsLinkComponent
Uridine, Millipore®

Uridine, Millipore®

Glass bottle for solids, 100 g
Catalog # 6680-100
Uridine, Millipore®
Catalog # 6680-100
CAS Number:  58-96-8
Uridine, Millipore®

Glass bottle for solids, 100 g

CAS Number:  58-96-8
Price
462,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.