Order Entry
Belgium
Orders LinkContactUsLinkComponent
2-Phenylethanol 99%

2-Phenylethanol 99%

Glass bottle, 1 L
Catalog # ACRO130180010
Irritant
2-Phenylethanol 99%
Catalog # ACRO130180010
CAS Number:  60-12-8
2-Phenylethanol 99%
2-Phenylethanol 99%

Glass bottle, 1 L

CAS Number:  60-12-8
Price
72,30 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
  • CAS Number:60-12-8
  • Formula:C₆H₅CH₂CH₂OH
  • MDL Number:MFCD00002886
  • Molecular Weight:122.17 g/mol
  • Merck Index:12,07370
  • Boiling Pt:219 °C (760 mmHg)
  • Melting Pt:–27 °C
  • Density:1.02 g/cm³
  • Flash Pt:102 °C
  • Storage Temperature:Ambient
  • EINECS:200-456-2
  • ADR:6.1,III

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5300 to 1.5350 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific gravity:
    (25/25°C) 1.016 to 1.021
  • UV:
    at 220 nm A: ≤0.04
  • Titration after Ion exchange:
    ≥99.0 % (on dry substance)
  • Membrane filtration test:
    (on 0.2 M soln.) Passes test
  • Suitability:
    (HPLC gradient) Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.