Order Entry
Belgium
Orders LinkContactUsLinkComponent
4-Fluorophenol 99%

4-Fluorophenol 99%

Glass bottle for solids, 100 g
Catalog # ACRO119561000
Irritant
4-Fluorophenol 99%
Catalog # ACRO119561000
CAS Number:  371-41-5
4-Fluorophenol 99%
4-Fluorophenol 99%

Glass bottle for solids, 100 g

CAS Number:  371-41-5
Price
105,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:44…49 °C
  • Density:1,22 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2811
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow to beige to pink Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    44 to 49 °C
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.