Order Entry
Belgium
Orders LinkContactUsLinkComponent
Accessories

Accessories

Conductivity flow cell, titanium/CTFE, 14 μl volume, max. flow rate 100 ml/min, 16" outer diameter
Catalog # 18-1111-05
Supplier:   Cytiva
Accessories
Catalog # 18-1111-05
Supplier:  Cytiva
Accessories
Accessories

Conductivity flow cell, titanium/CTFE, 14 μl volume, max. flow rate 100 ml/min, 16" outer diameter

Price
1 990,00 €
Log in for availability
1 * 1 ST
Specifications
  • Description:
    Conductivity flow cell, titanium/CTFE, 14 μl volume, max. flow rate 100 ml/min, 16" outer diameter
  • Cat. No.:
    18-1111-05
  • W×D×H:
    260×370×100 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.