Orders LinkContactUsLinkComponent
Soil hydrometers
Soil hydrometers
Soil hydrometer, 0,995 − 1,038 S.G., 0,001 S.G., 280 mm
Catalog # 614-1534
Supplier:   Alla France
Soil hydrometers
Catalog # 614-1534
Supplier:  Alla France
Soil hydrometers
Soil hydrometers
Soil hydrometer, 0,995 − 1,038 S.G., 0,001 S.G., 280 mm
Price
83,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Soil hydrometers for specific gravity (SG) measurements.
  • Medium surface tension (55 mN/m)
  • Reading below meniscus
Certifications: ASTM E100 D422 - A2LA traceable.
Ordering information: EU traceable calibration certificate or EO official calibration certificate available on request.
Specifications
  • Description:
    Soil hydrometer
  • Length:
    280 mm
  • Seq:
    01
  • Division:
    0,001 S.G.
  • Range:
    0,995 − 1,038 S.G.