Broad range hydrometers
1,000 − 2,000 °Bé, 0,01, 300 mm
Catalog # 765-0052
Supplier: Alla France
Jump to:
Product Details & Documents
These broad range hydrometers are calibrated at 20 °C.
Mostly dedicated to applications with citric and tartaric acids.
Specifications
- Length:300 mm
- Division:0,01
- Range:1,000 − 2,000 °Bé



