Vanadium(V) oxide ≥99.2%
Plastic bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.2% CAS No.: 1314-62-1 Molecular Formula: V2O5 Formula Weight: 181.88 Physical Form: Powder Density: 3.357 Melting Point: 690° Boiling Point: 1750° dec. Solubility: Slightly soluble in water. Soluble in acids and alkalies Merck Reference: 14,9921 UN No.: 2862 MDL No.: MFCD00011457
As an oxidation catalyst, in refractories, as a ceramics Colourant, in the manufacture of vanadium salts, and as a dye
Synonyms: Vanadium pentoxide, di-Vanadium pentaoxide
- CAS Number:1314-62-1
- Formula:O₅V₂
- MDL Number:MFCD00011457
- Molecular Weight:181.88 g/mol
- Merck Index:14,9921
- Boiling Pt:1750 °C (dec.)
- Melting Pt:690 °C
- Density:3.357
- Storage Temperature:Ambient
- EINECS:215-239-8
- UN:2862
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g



