Order Entry
Belgium
Orders LinkContactUsLinkComponent
Accessories for fluid and solid samplers

Accessories for fluid and solid samplers

Suction hose, PTFE, Ø: 6 mm, thickness = 1 mm
Catalog # 228-1190
Supplier:   BURKLE
Accessories for fluid and solid samplers
Catalog # 228-1190
Supplier:  BURKLE
Accessories for fluid and solid samplers
Accessories for fluid and solid samplers

Suction hose, PTFE, Ø: 6 mm, thickness = 1 mm

Price
85,60 €
Log in for availability
1 * 5 m
Product Details & Documents
Specifications
  • Description:
    Suction hose, PTFE, Ø: 6 mm, thickness = 1 mm
  • Seq:
    12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.