Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Boxes, Multipurpose

Storage Boxes, Multipurpose

29 l, White, 660×450×130 mm, HDPE
Catalog # 216-1720
Supplier:   BURKLE
Storage Boxes, Multipurpose
Catalog # 216-1720
Supplier:  BURKLE
Storage Boxes, Multipurpose
Storage Boxes, Multipurpose

29 l, White, 660×450×130 mm, HDPE

Price
70,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of HDPE, these practical storage containers are ideal for safe transport of pourable or liquid contents.

  • With closed gripping edges
  • Can be stacked
  • Rounded inner edges and smooth inner surfaces for easy cleaning
  • Enables use in foodstuffs/hygiene sector
Specifications
  • Capacity:
    29 l
  • Colour:
    White
  • Material:
    HDPE
  • Seq:
    01
  • W×D×H:
    660×450×130 mm
Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Lid for universal storage containers, PS, white
    Lid for universal storage containers, PS, white
    BURK3418-0001
    - mm
    Log in for availability
    1 * 1 ST - 39,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.