Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sample dippers

Sample dippers

Dip sampler for liquid samples, Transparent, PP, 500 mm, 22 mm
Catalog # BURK5315-0050
Supplier:   BURKLE
Sample dippers
Catalog # BURK5315-0050
Supplier:  BURKLE
Sample dippers
Sample dippers

Dip sampler for liquid samples, Transparent, PP, 500 mm, 22 mm

Price
82,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Plunging siphons for the simple removal of liquid samples.

  • Made from transparent PP
  • All-layer and point sampling
  • Upper end with reflux valve, lower end with ball valve
Specifications
  • Ø:
    22 mm
  • Colour:
    Transparent
  • Description:
    Dip sampler for liquid samples
  • Length:
    500 mm
  • Material:
    PP
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.